Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodifluoroacetic anhydride

Catalog No. 25206925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-206-925G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-206-925G Supplier Sigma Aldrich Supplier No. 25206925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorodifluoroacetic anhydride

Specifications

CAS 2834-23-3
Density 1.395 g/mL (at 25°C (literature))
Boiling Point 96°C to 97°C (lit.)
Molecular Formula C4Cl2F4O3
Refractive Index n20/D 1.347 (literature)
Linear Formula (ClCF2CO)2 O
MDL Number MFCD00000774
Quantity 100 mg
Molecular Weight (g/mol) 242.94
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.