Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodiisopropylphosphine

Catalog No. 33777325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-777-325G 25 g
33-777-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 33-777-325G Supplier Sigma Aldrich Supplier No. 33777325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorodiisopropylphosphine can be used: To synthesize p-styryldiisopropylphosphine by reacting with 4-chlorostyrene via Grignard reaction.As a phosphination reagent in combination with [Cp2Zr(1-butene)(DMAP)] (Cp=cyclopentadienyl; DMAP= 4-(dimethylamino)pyridine)) for the zirconophosphination of alkynes to form zirconoalkenylphosphines.A luminescent mixed-donor platinum POCN pincer complex via cyclometalation process.

Specifications

CAS 40244-90-4
Density 0.959 g/mL at 25°C
Boiling Point 69°C (33 mmHg, lit.)
Molecular Formula C6H14ClP
Refractive Index n20/D 1.475
Linear Formula [(CH3)2CH]2PCl
MDL Number MFCD00015027
Quantity 25 g
Molecular Weight (g/mol) 152.6 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.