Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorodiisopropylsilane, 95%

Catalog No. AC313440050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC313440050 Supplier Thermo Scientific Chemicals Supplier No. 313440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2227-29-4 | C6H14ClSi | 149.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2227-29-4
Molecular Formula C6H14ClSi
Molecular Weight (g/mol) 149.71
MDL Number MFCD00054896
InChI Key IGSUJBNDAWQLST-UHFFFAOYSA-N
Synonym chlorodiisopropylsilane, diisopropylchlorosilane, unii-dvg177547j, silane, chloro diisopropyl, zlchem 966, chlorodiisopropylsilyl, chlorobis propan-2-yl silane, chloro-di propan-2-yl silicon
PubChem CID 6365034
IUPAC Name chloro-di(propan-2-yl)silicon
SMILES CC(C)[Si](Cl)C(C)C

Specifications

Color Colorless
Density 0.8720g/mL
Boiling Point 137.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.872
Merck Index 15,2136
Solubility Information Solubility in water: reacts
Formula Weight 150.73
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material chlorodiisopropylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Handle under inert gas.
Protect from moisture.
Keep away from heat/spa

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.