Learn More
Medchemexpress LLC Chlorodimethoxytriazine | 3140-73-6 | MFCD00075607 | 98.9% | 175.57 g/mol | C5H6ClN3O2 | 500 G
SDP
Catalog No. 5000303781
Encompass_Preferred
Chlorodimethoxytriazine (2-chloro-4,6-dimethoxy-1,3,5-triazine, CAS 3140-73-6) is an organic biochemical reagent used in organic synthesis and peptide coupling applications. It is supplied as a purified crystalline powder with documented assay and supporting analytical and safety documents for laboratory research use.
  • Purity: 98.9% assay reported on product documentation
  • White crystalline powder, suitable for accurate weighing
  • Useful as a coupling reagent and building block in organic synthesis
  • Certificate of analysis and safety data sheet available for verification
  • Available in multiple laboratory pack sizes to suit research needs
  • For research use only; not for clinical or human use
Catalog No. 50-003-03781 Supplier Medchemexpress LLC Supplier No. HYW016718500G
Chlorodimethoxytriazine (2-chloro-4,6-dimethoxy-1,3,5-triazine, CAS 3140-73-6) is an organic biochemical reagent used in organic synthesis and peptide coupling applications. It is supplied as a purified crystalline powder with documented assay and supporting analytical and safety documents for laboratory research use.
  • Purity: 98.9% assay reported on product documentation
  • White crystalline powder, suitable for accurate weighing
  • Useful as a coupling reagent and building block in organic synthesis
  • Certificate of analysis and safety data sheet available for verification
  • Available in multiple laboratory pack sizes to suit research needs
  • For research use only; not for clinical or human use