Learn More
Chlorodimethylphenylsilane, 97+%
Description
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.
Solubility
Miscible with tetrahydrofuran, ether and hexane.
Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
Specifications
Specifications
| Boiling Point | 192°C to 193°C |
| Quantity | 5 g |
| UN Number | UN2987 |
| Beilstein | 606292 |
| Flash Point | 72°C (161°F) |
| Solubility Information | Miscible with tetrahydrofuran,ether and hexane. |
| Formula Weight | 170.72 |
| Percent Purity | ≥97% |
| Refractive Index | 1.509 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H227-H314 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| EINECSNumber | 212-193-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.