Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodiphenylphosphine

Catalog No. 503119099 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-9099 100 g
50-311-9100 25 g
50-311-9130 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 50-311-9099 Supplier Sigma Aldrich Supplier No. C39601100G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to introduce the diphenylphosphinyl moiety by aryl ortho-lithiation.

Specifications

CAS 1079-66-9
Density 1.194 g/mL at 25°C, 1.229 g/mL at 25°C
Boiling Point 320°C
Molecular Formula C12H10ClP
Refractive Index n20/D 1.636
Linear Formula (C6H5)2PCl
MDL Number MFCD00000529
Quantity 100 g
Synonym P-Chlorodiphenylphosphine; ClPPh2; PPh2Cl; Diphenylphosphinous chloride
Molecular Weight (g/mol) 220.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.