Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodiphenylphosphine

Catalog No. 503119130 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-9130 500 g
50-311-9099 100 g
50-311-9100 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 50-311-9130 Supplier Sigma Aldrich Supplier No. C39601500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to introduce the diphenylphosphinyl moiety by aryl ortho-lithiation.

Specifications

CAS 124-83-4
Melting Point 183°C to 186°C
Molecular Formula C10H16O4
Linear Formula C10H16O4
Refractive Index n20/D 1.636
MDL Number MFCD00001375
Quantity 500 g
Synonym P-Chlorodiphenylphosphine; ClPPh2; PPh2Cl; Diphenylphosphinous chloride
Molecular Weight (g/mol) 200.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.