Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodiphenylphosphine 97.0+%, TCI America™
SDP

Catalog No. C0597100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0597100G 100 g
C059725G 25 g
C0597500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0597100G Supplier TCI America Supplier No. C0597100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
IUPAC Name chlorodiphenylphosphane
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 215°C
Quantity 100 g
UN Number 3265
Formula Weight 220.64
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
EINECSNumber (3)-3943
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.