Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroethane (ca. 17% in Ethyl Ether, ca. 2.0mol/L), TCI America™
SDP

Catalog No. C2882100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2882100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2882100ML Supplier TCI America Supplier No. C2882100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-00-3
Molecular Formula C2H5Cl
Molecular Weight (g/mol) 64.512
MDL Number MFCD00000961
InChI Key HRYZWHHZPQKTII-UHFFFAOYSA-N
Synonym ethyl chloride, ethane, chloro, chlorethyl, monochloroethane, chlorene, chelen, muriatic ether, monochlorethane, anodynon, cloretilo
PubChem CID 6337
ChEBI CHEBI:47554
IUPAC Name chloroethane
SMILES CCCl

Specifications

Color Colorless
Quantity 100 mL
UN Number 1993
Formula Weight 64.51
Physical Form Liquid
Chemical Name or Material Chloroethane (ca. 17% in Ethyl Ether, ca. 2.0mol/L)
RTECSNumber KH7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.