Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chloroethene (polymer) | 9002-86-2 | MFCD00084459 | 20,000-80,000 | (C2H3Cl)x | 50 G
SDP

Catalog No. 5000284050
Encompass_Preferred

Chloroethene (polymer), also known as polyvinyl chloride (PVC), is supplied as a white to off-white powder for research and analytical applications. It has an estimated molecular weight range of 20,000-80,000 and is used to study packaging inner surfaces, food wrap, agricultural coverings, and plastic bottle materials.

  • White to off-white powder suitable for analytical research.
  • Molecular formula (C2H3Cl)x with Mw range 20,000-80,000.
  • Packaged as 50 g for laboratory use and handling.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Intended for research on packaging materials, food contact films, and agricultural coverings.

Catalog No. 50-002-84050 Supplier Medchemexpress LLC Supplier No. HYW687022A50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chloroethene (polymer), also known as polyvinyl chloride (PVC), is supplied as a white to off-white powder for research and analytical applications. It has an estimated molecular weight range of 20,000-80,000 and is used to study packaging inner surfaces, food wrap, agricultural coverings, and plastic bottle materials.

  • White to off-white powder suitable for analytical research.
  • Molecular formula (C2H3Cl)x with Mw range 20,000-80,000.
  • Packaged as 50 g for laboratory use and handling.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Intended for research on packaging materials, food contact films, and agricultural coverings.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.