Learn More
Chloroform, 200 μg/mL Analytical Standard, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 48603
Description
Specifications
| 1 mL | |
| Ampule | |
| UN1230 - Class 3 - PG 2 - Methanol, solution | |
| 200 μg/mL in methanol |
| 11°C (Closed Cup) | |
| MFCD00000826 | |
| Analytical Standard |
Safety and Handling
H225 - H301 + H311 + H331 - H370
Recommended Storage : 2°C to 8°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.