Learn More
Andwin Scientific BDH Chloroform BDH 4L Glass BDH Chloroform
SDP
Catalog No. NC9011194
Encompass_Preferred
BDH Chloroform BDH 4L Glass BDH Chloroform (With Amylene Preservative) Is Suitable For General Laboratory Use Including Cytology Histology Organic Synthesis Cleaning Prep Scale Synthesis. Packaged In 4-Liter Glass Bottles With 4 Bottles Per Case. ACS Ce
Catalog No. NC9011194 Supplier Andwin Scientific Supplier No. BDH11094LG
BDH Chloroform BDH 4L Glass BDH Chloroform (With Amylene Preservative) Is Suitable For General Laboratory Use Including Cytology Histology Organic Synthesis Cleaning Prep Scale Synthesis. Packaged In 4-Liter Glass Bottles With 4 Bottles Per Case. ACS Ce