Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Chloroform, 500 mL

Catalog No. 501995032 Shop All Hach Company Products
Encompass

ACS Grade, CAS No. 67-66-3 CHCl3, FW = 119.38.Assay: ≥99.8% 500 mL

Catalog No. 50-199-5032 Supplier Hach Company Supplier No. 1445849
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Grade, CAS No. 67-66-3 CHCl3, FW = 119.38.Assay: ≥99.8% 500 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.