Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform contains 100-20
Catalog No. 502899040
Encompass
Chloroform is a colorless volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.
Catalog No. 50-289-9040 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C24322.5L
Chloroform is a colorless volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.