Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform contains 100-20
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C24324X25ML
Encompass
Chloroform is a colorless volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.
Catalog No. 50-289-7074