Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform contans 100 20500ML
Catalog No. 502903632
Encompass
Chloroform is a colorless volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.
Catalog No. 50-290-3632 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C24326X500ML
Chloroform is a colorless volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.