Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform-d 99.8 atom % D, contains 0.03 % (v/v) TMS | 865-49-6 | MFCD00000827 | 100G
Catalog No. NC2176818
Encompass
Chloroform-d 99.8 atom % D, contains 0.03 % (v/v) TMS | Purity: >=99% (CP) | Mol Wt: 120.38 | 865-49-6 | MFCD00000827 | 100G
Catalog No. NC2176818 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 225789100G
Chloroform-d 99.8 atom % D, contains 0.03 % (v/v) TMS | Purity: >=99% (CP) | Mol Wt: 120.38 | 865-49-6 | MFCD00000827 | 100G