Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform-d >=99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer | 865-49-6 | MFCD00000827 | 250G
Catalog No. 501859886
Encompass
Chloroform-d >=99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer | Purity: >=99.00% | Mol Wt: 120.38 | 865-49-6 | MFCD00000827 | 250G
Catalog No. 50-185-9886 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 416754250G
Chloroform-d >=99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer | Purity: >=99.00% | Mol Wt: 120.38 | 865-49-6 | MFCD00000827 | 250G