Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform-d, 99.8% (Isotopic), contains 0.03% v/v TMS

Catalog No. AA4138914
Change view
Click to view available options
Quantity:
25 g
25 mL
100 g
5 x 100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4138914 25 g
AA41389AC 25 mL
AA4138922 100 g
AA4138959 5 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4138914 Supplier Thermo Scientific Chemicals Supplier No. 04138914
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 865-49-6 | CHCl3 | 120.375 g/mol

Widely used in the organic solvent for the NMR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.375
MDL Number MFCD00000827
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
Synonym chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
PubChem CID 71583
ChEBI CHEBI:85365
IUPAC Name trichloro(deuterio)methane
SMILES C(Cl)(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Sensitivity Light and Moisture
Quantity 25 g
Density 1.50 g/mL
Refractive Index 1.4445
Melting Point -64°C
Boiling Point 62°C
UN Number UN1888
Solubility Information Miscible with organic solvents,ethyl acetate and acetone.
Formula Weight 120.38
Percent Purity 99.8%
Concentration or Composition (by Analyte or Components) contains 0.03% v/v TMS
Chemical Name or Material Chloroform-d, Isotopic
Show More Show Less
Hazard Category H302-H315-H319-H331-H336-H351-H361d-H372
Hazard Statement GHS H Statement
H351-H373-H302-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROFORM
EINECSNumber 212-742-4
TSCA No
storageNote1 2°C to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.