Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform-d, for NMR, 100.0 atom % D, stabilized with silver

Catalog No. AC464050250
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464050250 25 mL
AC464051000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464050250 Supplier Thermo Scientific Chemicals Supplier No. 464050250
Add to Cart

CAS: 865-49-6 | CHCl3 | 120.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.38
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
IUPAC Name trichloro(²H)methane
SMILES [2H]C(Cl)(Cl)Cl

Specifications

Density 1.5000 g/mL
Specific Gravity 1.5
Quantity 25 mL
Melting Point -64.0°C
Boiling Point 60.0°C
Linear Formula CDCl3
Solubility Information Solubility aq. soln.: 8.2 g/L (20°C).
Formula Weight 120.39
Grade NMR
Chemical Name or Material Chloroform-d
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed.

Toxic if inhaled.

Causes skin irritation.

Causes serious eye irritation.

Suspected of causing cancer.

Causes damage to organs through prolonged or repeated exposure.

May cau
Precautionary Statement GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician.

IF ON SKIN: Wash with plenty of soap and water.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 212-742-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.