Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform-d, for NMR, 100 atom % D, packaged in 0.75 ml ampoules

Catalog No. AC320680075
Change view
Click to view available options
Quantity:
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC320680075 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC320680075 Supplier Thermo Scientific Chemicals Supplier No. 320680075
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 865-49-6 | CHCl3 | 120.375 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.375
MDL Number MFCD00000827
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
Synonym chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
PubChem CID 71583
ChEBI CHEBI:85365
IUPAC Name trichloro(deuterio)methane
SMILES C(Cl)(Cl)Cl

Specifications

Physical Form Liquid
Quantity 7.5 mL
Color Colorless
Deuterium (D) 99.95% min.
Density 1.5000g/mL
Water 0.01% max. (Coulometric)
Melting Point -64.0°C
Boiling Point 60.0°C
Linear Formula CDCl3
UN Number 1888
Beilstein 01, IV, 54
Formula Weight 120.39
Grade NMR
Chemical Name or Material Chloroform-d, 100 atom % D
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.