Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR

Specifications
Specifications
Physical Form | Crystalline Powder |
Density | 1.5000g/mL |
Color | White to Beige |
Packaging | Glass Bottle |
Quantity | 25 mL |
Deuterium (D) | 99.80% min. |
Water | 0.01% max. (Coulometric) |
Melting Point | -64.0°C |
Boiling Point | 60.0°C |
Linear Formula | CDCl3 |
Show More |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child. |
Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. |
EINECSNumber | 212-742-4 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction