Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform - D1, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv™, MilliporeSigma™

Catalog No. M1024500100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1024500100 100 mL Glass Bottle
M1024500500 500 mL Glass Bottle
M1024500025 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1024500100 Supplier MilliporeSigma Supplier No. 1.02450.0100
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.375
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
Synonym chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
PubChem CID 71583
ChEBI CHEBI:85365
IUPAC Name trichloro(deuterio)methane
SMILES C(Cl)(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 100 mL
Vapor Pressure 211 hPa at 20°C (68°F)
Density 1.50g/cm3 at 20°C (68°F)
Odor Characteristic
Packaging Glass Bottle
Melting Point -64.1°C
Boiling Point 60°C
Assay Percent Range 99.8% min.
UN Number 1888
Formula Weight 120.38
Chemical Name or Material Chloroform - D1, Stabilized with silver
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.