Learn More
Crescent Chemical Co Inc Chloroform:Isoamyl Alcohol 24:1, 500mL
SDP
Catalog No. 50248380
Encompass_Preferred
Molecular Biology Grade, DNase, RNase, Proteases not detected. Mixture of chloroform and isoamyl alcohol (ratio 24:1). Bottled under inert gas. Suitable for nucleic acid purification. Improves efficiency of nucleic acid extraction. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-380 Supplier Crescent Chemical Co Inc Supplier No. SE39554.02
Molecular Biology Grade, DNase, RNase, Proteases not detected. Mixture of chloroform and isoamyl alcohol (ratio 24:1). Bottled under inert gas. Suitable for nucleic acid purification. Improves efficiency of nucleic acid extraction. Storage Conditions: +15 °C TO +30 °C