Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform Isoamyl Alcohol Mixture, BioUltra, For molecular biology, 24:1, 99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018224 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8224 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8224 Supplier MilliporeSigma Supelco Supplier No. 25666500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Quantity 500 mL
MDL Number MFCD00171265
Percent Purity ≥99.5% (chloroform + isoamyl alcohol, GC)
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.