Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform/Methanol for Karl Fischer, Reagents

Catalog No. 01335674
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-674 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-674 Supplier Reagents Holdings Llc Supplier No. CC1010004C
Add to Cart
Add to Cart

Chloroform/Methanol for Karl Fischer, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Color Colorless
CAS 67-56-1, 67-66-3
Physical Form Liquid
Quantity 4 L
Packaging Glass Bottle
Specific Gravity 1.3343
Grade Reagent
Chemical Name or Material Chloroform/Methanol for Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.