Learn More
Crescent Chemical Co Inc Chloroform, 67-66-3, MFCD00000826, 250mL
SDP
Catalog No. 50248378
Encompass_Preferred
Molecular Formula CHCl3, Molecular Weight 119.378, Molecular Biology Grade, Suitable for nucleic acid purification and for removal of traces of phenol from aqueous DNA and RNA samples. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-378 Supplier Crescent Chemical Co Inc Supplier No. SE39553.01
Molecular Formula CHCl3, Molecular Weight 119.378, Molecular Biology Grade, Suitable for nucleic acid purification and for removal of traces of phenol from aqueous DNA and RNA samples. Storage Conditions: +15 °C TO +30 °C