Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform ReagentPlus(R), >=99.8%, contains 0.5-1.0% ethanol as stabilizer | 67-66-3 | MFCD00000826 | 2.5L
Catalog No. 501884217
Encompass
Chloroform ReagentPlus(R), >=99.8%, contains 0.5-1.0% ethanol as stabilizer | Purity: >=99.8% | Mol Wt: 119.38 | 67-66-3 | MFCD00000826 | 2.5L
Catalog No. 50-188-4217 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1329502.5L
Chloroform ReagentPlus(R), >=99.8%, contains 0.5-1.0% ethanol as stabilizer | Purity: >=99.8% | Mol Wt: 119.38 | 67-66-3 | MFCD00000826 | 2.5L