Learn More
Cayman Chemical Chlorogenic Acid, Purity: ≥95%, CAS: 327-97-9, 500 mg
SDP
Catalog No. 502896783
Encompass_Preferred
Chlorogenic acid is a phenolic natural product isolated from the leaves and fruits of dicotyledonous plants, including the coffee bean. Structurally, chlorogenic acid is the ester of caffeic acid with the 3-hydroxyl group of quinic acid. Chlorogenic acid is an important factor in plant metabolism. It is also an antioxidant and an inhibitor of the tumor promoting activity of phorbol esters.{1430,1420} Chlorogenic acid, at concentrations as high as 100 µM, does not inhibit the 5-LO activity of ionophore-stimulated human PMNL.{4706}
Catalog No. 50-289-6783 Supplier Cayman Chemical Supplier No. 70930500 MG
Chlorogenic acid is a phenolic natural product isolated from the leaves and fruits of dicotyledonous plants, including the coffee bean. Structurally, chlorogenic acid is the ester of caffeic acid with the 3-hydroxyl group of quinic acid. Chlorogenic acid is an important factor in plant metabolism. It is also an antioxidant and an inhibitor of the tumor promoting activity of phorbol esters.{1430,1420} Chlorogenic acid, at concentrations as high as 100 µM, does not inhibit the 5-LO activity of ionophore-stimulated human PMNL.{4706}