Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chlorogenic acid, 98%

Catalog No. AC109240050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109240050 5 g Glass bottle
AC109240010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109240050 Supplier Thermo Scientific Chemicals Supplier No. 109240050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 327-97-9 | C16H18O9 | 354.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16H18O9
Molecular Weight (g/mol) 354.31
MDL Number MFCD00003862
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
PubChem CID 1794427
ChEBI CHEBI:16112
IUPAC Name (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O

Specifications

Melting Point 205.0°C to 209.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 10, I, 271
Merck Index 15, 2143
Specific Rotation -36
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and acetone
Formula Weight 354.3
Percent Purity 98%
Physical Form Crystals, Powder, Needles or Chunks
Chemical Name or Material Chlorogenic acid, Predominantly trans, From coffee seeds
Show More Show Less
EINECSNumber 206-325-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.