Learn More
Medchemexpress LLC Chlorogenic acid butyl ester | 132741-56-1 | 410.42 | 1 MG
SDP
Catalog No. 5000416945
Encompass_Preferred
Chlorogenic acid butyl ester is a caffeoylquinic acid known for its potent melanogenesis inhibitory and antioxidant activities. It functions by inhibiting the expression of key factors involved in melanogenesis, including microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2.
  • Acts as a potent melanogenesis inhibitor.
  • Inhibits the expression of MITF, tyrosinase, TRP-1, and TRP-2.
  • Exhibits antioxidant activity.
Catalog No. 50-004-16945 Supplier Medchemexpress LLC Supplier No. HYN100161MG
Chlorogenic acid butyl ester is a caffeoylquinic acid known for its potent melanogenesis inhibitory and antioxidant activities. It functions by inhibiting the expression of key factors involved in melanogenesis, including microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2.
  • Acts as a potent melanogenesis inhibitor.
  • Inhibits the expression of MITF, tyrosinase, TRP-1, and TRP-2.
  • Exhibits antioxidant activity.