Learn More
Medchemexpress LLC Chlorogenic acid butyl ester | 132741-56-1 | 410.42 | 5 MG
SDP
Catalog No. 5000416852
Encompass_Preferred
Chlorogenic acid butyl ester is a caffeoylquinic acid and a potent melanogenesis inhibitor. It inhibits the expression of microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2. It also exhibits antioxidant activity.
  • Potent melanogenesis inhibitor
  • Inhibits MITF, tyrosinase, TRP-1, and TRP-2 expression
  • Shows antioxidant activity
  • Exhibits inhibitory activities with 33-62% reduction of melanin content at 100 μM concentration on a-MSH-stimulated B16 melanoma cells with no or almost no toxicity
Catalog No. 50-004-16852 Supplier Medchemexpress LLC Supplier No. HYN100165MG
Chlorogenic acid butyl ester is a caffeoylquinic acid and a potent melanogenesis inhibitor. It inhibits the expression of microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2. It also exhibits antioxidant activity.
  • Potent melanogenesis inhibitor
  • Inhibits MITF, tyrosinase, TRP-1, and TRP-2 expression
  • Shows antioxidant activity
  • Exhibits inhibitory activities with 33-62% reduction of melanin content at 100 μM concentration on a-MSH-stimulated B16 melanoma cells with no or almost no toxicity