Learn More
Medchemexpress LLC Chlorogenic acid butyl ester | 132741-56-1 | 410.42 | 50 MG
SDP
Catalog No. 5000416535
Encompass_Preferred
Chlorogenic acid butyl ester, a caffeoylquinic acid, is a potent melanogenesis inhibitor. It inhibits the expression of microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2. This compound also demonstrates antioxidant activity.
  • Caffeoylquinic acid
  • Exhibits inhibitory activities with 33-62% reduction of melanin content at 100 μM concentration on a-MSH-stimulated B16 melanoma cells
  • Potent melanogenesis inhibitor
  • Inhibits expression of MITF, tyrosinase, TRP-1, and TRP-2
  • Shows antioxidant activity
Catalog No. 50-004-16535 Supplier Medchemexpress LLC Supplier No. HYN1001650MG
Chlorogenic acid butyl ester, a caffeoylquinic acid, is a potent melanogenesis inhibitor. It inhibits the expression of microphtalmia-associated transcription factor (MITF), tyrosinase, tyrosinerelated protein 1 (TRP-1), and TRP-2. This compound also demonstrates antioxidant activity.
  • Caffeoylquinic acid
  • Exhibits inhibitory activities with 33-62% reduction of melanin content at 100 μM concentration on a-MSH-stimulated B16 melanoma cells
  • Potent melanogenesis inhibitor
  • Inhibits expression of MITF, tyrosinase, TRP-1, and TRP-2
  • Shows antioxidant activity