Learn More
Medchemexpress LLC Chlorogenic acid butyl ester | 132741-56-1 | 98.2% | 410.4 g/mol | C20H26O9 | 10 MG
SDP
Catalog No. 5000296094
Encompass_Preferred
Chlorogenic acid butyl ester is a caffeoylquinic acid derivative that functions as a melanogenesis inhibitor and exhibits antioxidant activity. It is supplied as a research-grade small molecule suitable for cellular assays investigating melanin synthesis and related enzyme expression.
  • Acts as a melanogenesis inhibitor by downregulating MITF, tyrosinase, TRP-1, and TRP-2.
  • Exhibits antioxidant activity.
  • High purity suitable for research applications.
  • Available in small pack sizes convenient for laboratory testing.
Catalog No. 50-002-96094 Supplier Medchemexpress LLC Supplier No. HYN1001610MG
Chlorogenic acid butyl ester is a caffeoylquinic acid derivative that functions as a melanogenesis inhibitor and exhibits antioxidant activity. It is supplied as a research-grade small molecule suitable for cellular assays investigating melanin synthesis and related enzyme expression.
  • Acts as a melanogenesis inhibitor by downregulating MITF, tyrosinase, TRP-1, and TRP-2.
  • Exhibits antioxidant activity.
  • High purity suitable for research applications.
  • Available in small pack sizes convenient for laboratory testing.