Learn More
AdipoGen Chloroguanabenz . acetate (5 mg)
SDP
Supplier:  AdipoGen AGMRC0036M005
Encompass_Preferred
Chemical. CAS: 23113-55-5. Formula: C8H7Cl3N4 . C2H4O2. MW: 265.5 . 60.1. Antiprion agent. Inhibitor of protein aggregation. Specifically reduces accumulation of a pathogenic fragment of Huntingtin in a transiently transfected cellular model of Huntington’s disease (HD). Tool for treating HD and other polyglutamine expansion associated diseases.
Catalog No. 50-205-4257