Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(4-Cyanophenyl)[™-1-[(S)-2-[Bis(4-Fluorophenyl]Phosphino]Ferrocenyl]Ethyldi-Tert-Butylphosphine]Nickel(II)

Catalog No. 900941250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-094-1250MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 90-094-1250MG Supplier Sigma Aldrich Supplier No. 900941250MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Amination of functionalized aryl chlorides with ammonia and ammonium salts.

Specifications

Melting Point 170°C
Molecular Formula C36H52N6O15S
Linear Formula C36H52N6O15S
Quantity 25 mg
Synonym PC biotin-NHS ester
Molecular Weight (g/mol) 840.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.