Learn More
Strem, An Ascensus Company CAS# 55102-19-7. 1g. Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct, 98%. MFCD04971968
SDP
Catalog No. 5090115359
Encompass
CAS# 55102-19-7. 1g. Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct, 98%. MFCD04971968. Molecular Weight: 924.39 (1016.53). Molecular Formula: RuHCl[P(C6H5)3]3∙CH3C6H5. Color/form: purple xtl. Strem# 93-4401. http://www.strem.com/catalog/v/93-4401/
Catalog No. 50-901-15359 Supplier Strem, An Ascensus Company Supplier No. 9344011G
CAS# 55102-19-7. 1g. Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct, 98%. MFCD04971968. Molecular Weight: 924.39 (1016.53). Molecular Formula: RuHCl[P(C6H5)3]3∙CH3C6H5. Color/form: purple xtl. Strem# 93-4401. http://www.strem.com/catalog/v/93-4401/