Learn More
Strem, An Ascensus Company CAS# 63936-85-6. 250mg. Chloromethyl(1,5-cyclooctadiene)palladium(II), 99%. MFCD16621448
SDP
Supplier:  Strem, An Ascensus Company 460255250MG
Encompass
CAS# 63936-85-6. 250mg. Chloromethyl(1,5-cyclooctadiene)palladium(II), 99%. MFCD16621448. Molecular Weight: 265.09. Molecular Formula: PdCl(CH3)(C8H12). Color/form: light grey pwdr. Strem# 46-0255. http://www.strem.com/catalog/v/46-0255/
Catalog No. 50-815-09