Learn More
Description
- Chloromethyl chloroformate was used as key reactant in the synthesis of:
- Novel aminocarbonyloxymethyl esters of diclofenac and flufenamic acid
- Highly water-soluble monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin a
- Series of 3-acyloxymethyloxycarbonyl-1-aryl-3-methyltriazenes
Specifications
Specifications
| CAS | 22128-62-7 |
| Density | 1.450 g/mL (at 20°C) |
| Quantity | 10 mL |
| Refractive Index | n20/D 1.428 |
| Molecular Formula | C2H2Cl2O2 |
| Linear Formula | ClCOOCH2Cl |
| MDL Number | MFCD00077688 |
| Boiling Point | 107°C to 108°C (lit.) |
| Molecular Weight (g/mol) | 128.94 |
| Percent Purity | ≥98.0% (GC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.