Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl chloroformate

Catalog No. 2318150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-181-50ML 50 mL
23-181-10ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-181-50ML Supplier Sigma Aldrich Supplier No. 2318150ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chloromethyl chloroformate was used as key reactant in the synthesis of:
  • Novel aminocarbonyloxymethyl esters of diclofenac and flufenamic acid
  • Highly water-soluble monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin a
  • Series of 3-acyloxymethyloxycarbonyl-1-aryl-3-methyltriazenes

Specifications

CAS 22128-62-7
Density 1.450 g/mL (at 20°C)
Quantity 50 mL
Refractive Index n20/D 1.428
Linear Formula ClCOOCH2Cl
MDL Number MFCD00077688
Boiling Point 107°C to 108°C (lit.)
Molecular Weight (g/mol) 128.94
Percent Purity ≥98.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.