Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl Ethyl Ether

Catalog No. 14267025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-267-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 14-267-025G Supplier Sigma Aldrich Supplier No. 14267025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloromethyl ethyl ether can be used to prepare:6-Benzyl-1-(ethoxymethyl)-5-iodopyrimidine-2,4(1H,3H)-dione, a key intermediate in the preparation of MKC-442 analog.Acylation catalyst for alcohols named 1,3-bis-[™-1-(2-naphthyl)ethyl]imidazoliumchloride by reacting with glyoxal-bis-[™-1-(2-naphthyl)ethyl]imine.

Specifications

CAS 135884-31-0
Melting Point 93°C to 98°C
Molecular Formula C9H14BNO4
Refractive Index n20/D 1.404
Linear Formula C9H14BNO4
MDL Number MFCD01318939
Quantity 25 g
Synonym N-Boc-2-pyrryl boronic acid
Molecular Weight (g/mol) 211.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.