Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Chloromethyl Menthyl Ether

Catalog No. 3499255G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-992-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-992-55G Supplier Sigma Aldrich Supplier No. 3499255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Chloromethyl menthyl ether can be used:As a starting material for the synthesis of 1-methyl-3-(+)-methylmenthoxide imidazolium chloride, a key intermediate for the preparation of Ag(I) N-heterocyclic carbene, applicable as a catalyst in the diboration of alkenes.As a substrate in the synthesis of commercially important O-substituted +a-methoxymethylketones.To determine enantiomeric excess (ee) of an iron chiral auxiliary named [Fe(CO)(++;5- C2H5)(PPh3)COCH3].

Specifications

CAS 607-01-2
Density 1.048 g/mL at 25°C
Boiling Point 293°C
Molecular Formula C14H15P
Refractive Index n20/D 1.467
Linear Formula (C6H5)2PCH2CH3
MDL Number MFCD00015170
Quantity 5 g
Molecular Weight (g/mol) 214.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.