Learn More
Medchemexpress LLC Chloromethyl methyl carbonate | 40510-81-4 | HY-43384 | 124.52 | 500 G
SDP
Catalog No. 5000404852
Encompass_Preferred
Chloromethyl methyl carbonate is a chemical compound primarily used as a laboratory chemical and in the manufacture of substances. It is classified as a highly flammable liquid and vapor, and can cause severe skin burns and eye damage. This compound is also harmful if swallowed.
  • Highly flammable liquid and vapor
  • Harmful if swallowed
  • Causes severe skin burns and eye damage
  • Suitable for laboratory research and substance manufacturing
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture and light
Catalog No. 50-004-04852 Supplier Medchemexpress LLC Supplier No. HY43384500G
Chloromethyl methyl carbonate is a chemical compound primarily used as a laboratory chemical and in the manufacture of substances. It is classified as a highly flammable liquid and vapor, and can cause severe skin burns and eye damage. This compound is also harmful if swallowed.
  • Highly flammable liquid and vapor
  • Harmful if swallowed
  • Causes severe skin burns and eye damage
  • Suitable for laboratory research and substance manufacturing
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture and light