Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl Methyl Ether

Catalog No. 10033125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-033-125G 25 g
10-033-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 10-033-125G Supplier Sigma Aldrich Supplier No. 10033125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloromethyl methyl ether (MOM-Cl) can be used as a reagent:For the introduction of the methoxymethyl (MOM) protecting group by converting alcohols to methoxy methyl (MOM) ethers.To prepare some methoxymethyl group containing metal complexes coordinated to carbonyl and dipyridyl ligands In the preparation of fluorene/chloromethyl methyl ether cross-linked polymers by Friedel-Crafts polymerization.

Specifications

CAS 107-30-2
Density 1.06 g/mL at 25°C
Boiling Point 55°C to 57°C
Molecular Formula C2H5ClO
Refractive Index n20/D 1.396
Linear Formula ClCH2OCH3
MDL Number MFCD00000885
Quantity 25 g
Synonym MOM chloride; Methoxymethyl chloride; Methyl chloromethyl ether
Molecular Weight (g/mol) 80.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.