Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl Methyl Sulfide

Catalog No. C5400725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C5400725G 25 g
C54007100G 100 g
50-311-9121 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. C5400725G Supplier Sigma Aldrich Supplier No. C5400725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylthiomethylating reagent for carbonyl and aromatic compounds. Methylene transfer reagent for iron(II) mediated cyclopropanation.

Specifications

CAS 2373-51-5
Density 1.153 g/mL at 25°C
Boiling Point 105°C
Molecular Formula C2H5ClS
Refractive Index n20/D 1.498
Linear Formula CH3SCH2Cl
MDL Number MFCD00000923
Quantity 25 g
Synonym Chlorodimethyl sulfide
Molecular Weight (g/mol) 96.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.