Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl octyl ether

Catalog No. 2945195G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-451-95G 5 g
29-451-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-451-95G Supplier Sigma Aldrich Supplier No. 2945195G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chloromethyl octyl ether has been employed as chloromethylation reagent:
  • In chloromethylation of calix[4]arene, methyl ethers of calix[6]arene and calix[8]arene
  • In the synthesis of chloromethylated poly(phthalazinone ether sulfone ketone)
  • In chloromethylation of polyetherethersulphone-polyethersulphone copolymers
  • For polymers

Specifications

CAS 24566-90-3
Density 0.924 g/mL (at 25°C (literature))
Molecular Formula C9H19ClO
Refractive Index n20/D 1.437 (literature)
Linear Formula CH3(CH2)7 OCH2Cl
MDL Number MFCD00010767
Quantity 5 g
Molecular Weight (g/mol) 178.7
Percent Purity 95%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.