Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl p-Tolyl Sulfide 96.0+%, TCI America™
SDP

Catalog No. C16015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16015G Supplier TCI America Supplier No. C16015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34125-84-3
Molecular Formula C8H9ClS
Molecular Weight (g/mol) 172.67
MDL Number MFCD01318451
InChI Key VFQYMJYUSRAJCZ-UHFFFAOYSA-N
Synonym 4-(Chloromethylthio)toluene, p-Tolyl Chloromethyl Sulfide
PubChem CID 1502059
IUPAC Name 1-[(chloromethyl)sulfanyl]-4-methylbenzene
SMILES CC1=CC=C(SCCl)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 172.67
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl p-Tolyl Sulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.