Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloromethyl Phenyl Sulfide

Catalog No. 21663125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-663-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-663-125G Supplier Sigma Aldrich Supplier No. 21663125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the high-yield thiomethylation of O-silylated lactone and ester enolates in the synthesis of +a-methylene lactones and esters.

Specifications

CAS 41864-22-6
Melting Point 138°C to 152°C
Molecular Formula C5H4N6O
Refractive Index n20/D 1.594
Linear Formula C5H4N6O
MDL Number MFCD00043399
Quantity 25 g
Synonym CDT
Molecular Weight (g/mol) 164.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.