Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyl Polystyrene Resin cross-linked with 2% DVB (100-200mesh) (0.8-1.2mmol/g), TCI America™
SDP

Catalog No. C17515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17515G Supplier TCI America Supplier No. C17515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55844-94-5
MDL Number MFCD00146406
Synonym Merrifield Resin

Specifications

Quantity 5 g
Chemical Name or Material Chloromethyl Polystyrene Resin cross-linked with 2% DVB (100-200mesh) (0.8-1.2mmol/g)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.