Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chloromethyl)trimethoxysilane 98.0+%, TCI America™
SDP

Catalog No. C30735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30735G 5 g
C307325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30735G Supplier TCI America Supplier No. C30735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5926-26-1
Molecular Formula C4H11ClO3Si
Molecular Weight (g/mol) 170.66
MDL Number MFCD00468107
InChI Key FPOSCXQHGOVVPD-UHFFFAOYSA-N
PubChem CID 22212
IUPAC Name (chloromethyl)trimethoxysilane
SMILES CO[Si](CCl)(OC)OC

Specifications

Color Colorless
Boiling Point 156°C
Quantity 5 g
UN Number 1993
Formula Weight 170.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Chloromethyl)trimethoxysilane
EINECSNumber (2)-2079
RTECSNumber VV2625000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.